Learn More
Grainger Crimp Fitting 45 Deg 3/4 I.D. M30-2 F
Catalog No. 502764537
Encompass_Preferred
The Grainger 16P832 is a 3/4 HP, 115V, single phase, open drip proof motor with a 1075 RPM operating speed. It has a 48Y frame with a 1.15 service factor and features a cast iron body with standard foot mounting. The motor is thermally protected and rated for continuous duty applications.
Catalog No. 50-276-4537 Supplier Grainger Supplier No. 16P832
The Grainger 16P832 is a 3/4 HP, 115V, single phase, open drip proof motor with a 1075 RPM operating speed. It has a 48Y frame with a 1.15 service factor and features a cast iron body with standard foot mounting. The motor is thermally protected and rated for continuous duty applications.