Learn More
Grainger Crimp Fitting 45 Deg 3/4 I.D. M36-2 F
Catalog No. 502764141
Encompass_Preferred
The Aeroquip 16P879 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and can be used with a wide variety of hydraulic tubing materials. This compact crimp fitting provides a reliable, heavy duty connection for hydraulic applications.
Catalog No. 50-276-4141 Supplier Grainger Supplier No. 16P870
The Aeroquip 16P879 hydraulic crimp fitting is made of steel and has a 1/4" NPTF thread size. It is designed for use with 1/4" OD tubing and has a working pressure of up to 5000 psi. The fitting has a hex nut for tightening and can be used with a wide variety of hydraulic tubing materials. This compact crimp fitting provides a reliable, heavy duty connection for hydraulic applications.