Learn More
Grainger Crimp Fitting 45 Deg 3/8 ID 5/8 M
Catalog No. 502752653
Encompass_Preferred
The Grainger Approved Crimp Fitting is a copper crimp fitting used to join copper tubing. It has a 1/4" NPT female thread and is made of wrought copper. The fitting accepts 1/4" OD tubing and is rated for a maximum pressure of 300 PSI at 250°F. It can be used for air, water, oil, natural gas, and steam applications. The fitting meets SAE J513 specifications.
Catalog No. 50-275-2653 Supplier Grainger Supplier No. 14L764
The Grainger Approved Crimp Fitting is a copper crimp fitting used to join copper tubing. It has a 1/4" NPT female thread and is made of wrought copper. The fitting accepts 1/4" OD tubing and is rated for a maximum pressure of 300 PSI at 250°F. It can be used for air, water, oil, natural gas, and steam applications. The fitting meets SAE J513 specifications.