Learn More
Grainger Crimp Fitting 45 Deg 3/8 ID M18-1.5 F
Catalog No. 502766407
Encompass_Preferred
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for high pressure hydraulic hose connections. It is made from carbon steel with zinc dichromate plating and is designed for use with 1-wire braid or 4-wire braid hose. This fitting has a 37 degree JIC 37° flare straight thread o-ring boss port and is available in sizes -04 through -16. It can handle pressures up to 5000 psi and temperatures up to 250°F. The crimp diameter is 1.25 inches.
Catalog No. 50-276-6407 Supplier Grainger Supplier No. 16P829
The Aeroquip Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for high pressure hydraulic hose connections. It is made from carbon steel with zinc dichromate plating and is designed for use with 1-wire braid or 4-wire braid hose. This fitting has a 37 degree JIC 37° flare straight thread o-ring boss port and is available in sizes -04 through -16. It can handle pressures up to 5000 psi and temperatures up to 250°F. The crimp diameter is 1.25 inches.