Learn More
Grainger Crimp Fitting 45 Deg 3/8 ID M16-1.5 F
Catalog No. 502765907
Encompass_Preferred
The Grainger website features a variety of crimp hydraulic hose fittings. These fittings are designed to connect hydraulic hoses in fluid power systems. They are available in different styles including straight, 45 degree, and 90 degree configurations. The fittings come in different sizes to accommodate various hose diameters. They are made from steel or stainless steel for durability. Key features include zinc plating for corrosion resistance and O-ring seals for leak-free connections. The fittings use hex crimp collars to securely attach to hydraulic hoses. Overall, these fittings provide reliable, heavy-duty connections for high-pressure hydraulic applications.
Catalog No. 50-276-5907 Supplier Grainger Supplier No. 16P828
The Grainger website features a variety of crimp hydraulic hose fittings. These fittings are designed to connect hydraulic hoses in fluid power systems. They are available in different styles including straight, 45 degree, and 90 degree configurations. The fittings come in different sizes to accommodate various hose diameters. They are made from steel or stainless steel for durability. Key features include zinc plating for corrosion resistance and O-ring seals for leak-free connections. The fittings use hex crimp collars to securely attach to hydraulic hoses. Overall, these fittings provide reliable, heavy-duty connections for high-pressure hydraulic applications.