Learn More
Grainger Crimp Fitting 45 Deg 3/8 ID M22-1.5 F
Catalog No. 502764566
Encompass_Preferred
The Grainger Approved Crimp Fitting is a copper crimp fitting used to join copper tubing. It has a 1/2" trade size and is made of wrought copper. The fitting has a pressure rating of 300 psi at 73°F and is NSF/ANSI 61 certified. It conforms to ASTM B75 and ASTM B88 standards. The crimp fitting is designed for use in potable water, oil, natural gas, steam, compressed air, and refrigerant systems. It requires a crimping tool to secure tubing. This copper crimp fitting provides a reliable, reusable fitting for copper tubing in a variety of applications.
Catalog No. 50-276-4566 Supplier Grainger Supplier No. 16P867
The Grainger Approved Crimp Fitting is a copper crimp fitting used to join copper tubing. It has a 1/2" trade size and is made of wrought copper. The fitting has a pressure rating of 300 psi at 73°F and is NSF/ANSI 61 certified. It conforms to ASTM B75 and ASTM B88 standards. The crimp fitting is designed for use in potable water, oil, natural gas, steam, compressed air, and refrigerant systems. It requires a crimping tool to secure tubing. This copper crimp fitting provides a reliable, reusable fitting for copper tubing in a variety of applications.