Learn More
Grainger Crimp Fitting 45 Deg 3/8 ID M20-1.5 F
Catalog No. 502763790
Encompass_Preferred
The Parker Hydraulic Crimp Fitting is made of steel and has a Parker Code 55CY93. It is a hydraulic crimp fitting that creates leak-proof connections. This fitting has a working pressure of 5000 psi and a temperature range of -65 to +250°F. It has a tube OD of 3/4 in, a thread size of 9/16-18 UNF-3A, and a tube wall thickness of 0.065 in. The Parker Hydraulic Crimp Fitting 55CY93 is designed for use in hydraulic systems to create secure, leak-free connections between tubing and hose assemblies.
Catalog No. 50-276-3790 Supplier Grainger Supplier No. 16P866
The Parker Hydraulic Crimp Fitting is made of steel and has a Parker Code 55CY93. It is a hydraulic crimp fitting that creates leak-proof connections. This fitting has a working pressure of 5000 psi and a temperature range of -65 to +250°F. It has a tube OD of 3/4 in, a thread size of 9/16-18 UNF-3A, and a tube wall thickness of 0.065 in. The Parker Hydraulic Crimp Fitting 55CY93 is designed for use in hydraulic systems to create secure, leak-free connections between tubing and hose assemblies.