Learn More
Grainger Crimp Fitting 45 Deg 5/8 ID 5/8 FBSPP
Catalog No. 502768425
Encompass_Preferred
The Grainger 16P790 is a 3M Scotch-Brite General Purpose Hand Pad with an open web construction and non-scratching fibers for cleaning and scouring. Measuring 6x9 inches, it has medium duty abrasiveness. Ideal for cleaning pots, pans, dishes, counters, walls, appliances, tools, and more, it removes grease, dirt, food residue, rust stains, soap scum, and grime without damaging surfaces. Usable wet or dry on stainless steel, ceramic, glass, plastic, chrome, and enamel, it is durable, lint-free, and rinses easily. This pad provides an effective scrubbing solution for light-duty cleaning tasks.
Catalog No. 50-276-8425 Supplier Grainger Supplier No. 16P790
The Grainger 16P790 is a 3M Scotch-Brite General Purpose Hand Pad with an open web construction and non-scratching fibers for cleaning and scouring. Measuring 6x9 inches, it has medium duty abrasiveness. Ideal for cleaning pots, pans, dishes, counters, walls, appliances, tools, and more, it removes grease, dirt, food residue, rust stains, soap scum, and grime without damaging surfaces. Usable wet or dry on stainless steel, ceramic, glass, plastic, chrome, and enamel, it is durable, lint-free, and rinses easily. This pad provides an effective scrubbing solution for light-duty cleaning tasks.