Learn More
Grainger Crimp Fitting 45 Deg 5/8 ID M26-1.5 F
Catalog No. 502766404
Encompass_Preferred
The Grainger 16P831 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor with a 1.15 Service Factor. It has a 48Y Frame with a 1" diameter shaft and operates on 60 Hz power. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and thermally protected for overload conditions. It can provide continuous duty power for pumps, fans, and other industrial applications requiring up to 3/4 horsepower.
Catalog No. 50-276-6404 Supplier Grainger Supplier No. 16P831
The Grainger 16P831 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor with a 1.15 Service Factor. It has a 48Y Frame with a 1" diameter shaft and operates on 60 Hz power. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and thermally protected for overload conditions. It can provide continuous duty power for pumps, fans, and other industrial applications requiring up to 3/4 horsepower.