Learn More
Grainger Crimp Fitting 5/8 I.D. M26-1.5 F Metric
Catalog No. 502765320
Encompass_Preferred
The Parker Hydraulic Crimp Fitting is made of steel and has a zinc finish. It is designed for use with hydraulic hoses and tubing. The fitting has a 37° JIC 37° flare straight thread o-ring boss port. It is rated for pressures up to 5000 psi and temperatures up to 250°F. The fitting has a hex size of 1-1/16 inches. It meets or exceeds SAE J514 specifications. This fitting allows easy assembly and installation with hydraulic hoses and tubing in fluid power systems.
Catalog No. 50-276-5320 Supplier Grainger Supplier No. 16P822
The Parker Hydraulic Crimp Fitting is made of steel and has a zinc finish. It is designed for use with hydraulic hoses and tubing. The fitting has a 37° JIC 37° flare straight thread o-ring boss port. It is rated for pressures up to 5000 psi and temperatures up to 250°F. The fitting has a hex size of 1-1/16 inches. It meets or exceeds SAE J514 specifications. This fitting allows easy assembly and installation with hydraulic hoses and tubing in fluid power systems.