Learn More
Grainger Crimp Fitting 5/8 I.D. M26-1.5 M Metric
Catalog No. 502763936
Encompass_Preferred
The Grainger 16P848 is a 3/4 HP, 115V, single phase, open drip proof motor with a 1075 RPM operating speed. It has a 48 frame size, features sleeve bearings, and is totally enclosed for protection against dust and light splashing. This motor can provide reliable performance for industrial equipment and machinery. With its 3/4 HP rating, it is suitable for powering pumps, fans, conveyors, compressors, and other shop tools that require up to 560 watts of power. The 1075 RPM operating speed allows efficient torque transmission. The 48 frame size enables compatibility with standard mounts. With proper installation and maintenance, this motor can have a long service life.
Catalog No. 50-276-3936 Supplier Grainger Supplier No. 16P848
The Grainger 16P848 is a 3/4 HP, 115V, single phase, open drip proof motor with a 1075 RPM operating speed. It has a 48 frame size, features sleeve bearings, and is totally enclosed for protection against dust and light splashing. This motor can provide reliable performance for industrial equipment and machinery. With its 3/4 HP rating, it is suitable for powering pumps, fans, conveyors, compressors, and other shop tools that require up to 560 watts of power. The 1075 RPM operating speed allows efficient torque transmission. The 48 frame size enables compatibility with standard mounts. With proper installation and maintenance, this motor can have a long service life.