Learn More
Grainger Crimp Fitting 5/8 I.D. M30-2 F Metric
Catalog No. 502764513
Encompass_Preferred
The Parker Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for high pressure hydraulic hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -65 to +250°F. The fitting body is made of carbon steel with a zinc dichromate finish. It uses a hex crimp design and is compatible with many Parker industrial hoses. The crimp fitting creates a strong, reusable fitting for hydraulic systems.
Catalog No. 50-276-4513 Supplier Grainger Supplier No. 16P860
The Parker Hydraulic Crimp Fitting is a steel hydraulic fitting that uses crimp technology for high pressure hydraulic hose connections. It has a working pressure of up to 5000 PSI and a temperature range of -65 to +250°F. The fitting body is made of carbon steel with a zinc dichromate finish. It uses a hex crimp design and is compatible with many Parker industrial hoses. The crimp fitting creates a strong, reusable fitting for hydraulic systems.