Learn More
Grainger Crimp Fitting 90 Deg 1-1/4 F BSPP
Catalog No. 502768427
Encompass_Preferred
The Grainger 16P802 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA 48Y frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and double sealed ball bearings. It can be used for general purpose applications and meets NEMA design B standards. The motor dimensions are 6-7/16" Diameter x 10-3/4" Length and it weighs 26 pounds.
Catalog No. 50-276-8427 Supplier Grainger Supplier No. 16P802
The Grainger 16P802 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA 48Y frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and double sealed ball bearings. It can be used for general purpose applications and meets NEMA design B standards. The motor dimensions are 6-7/16" Diameter x 10-3/4" Length and it weighs 26 pounds.