Learn More
Grainger Crimp Fitting 90 Deg 1/2 ID 5/8 FBSPP
Catalog No. 502766736
Encompass_Preferred
The 1/2" Crimp Fitting, 90 Degree, 1/2" ID x 5/8" FBSPP is a brass compression fitting with a 90 degree angle. It has a 1/2" ID and 5/8" FBSPP threads. The fitting allows you to make a 90 degree turn in tubing runs. It is made of brass and is designed for use with soft copper tubing. This fitting requires a crimp ring (sold separately) to secure the tubing. It can handle pressures up to 850 psi and temperatures up to 250°F. This fitting is commonly used in hydraulic, pneumatic, and industrial plumbing applications.
Catalog No. 50-276-6736 Supplier Grainger Supplier No. 16P797
The 1/2" Crimp Fitting, 90 Degree, 1/2" ID x 5/8" FBSPP is a brass compression fitting with a 90 degree angle. It has a 1/2" ID and 5/8" FBSPP threads. The fitting allows you to make a 90 degree turn in tubing runs. It is made of brass and is designed for use with soft copper tubing. This fitting requires a crimp ring (sold separately) to secure the tubing. It can handle pressures up to 850 psi and temperatures up to 250°F. This fitting is commonly used in hydraulic, pneumatic, and industrial plumbing applications.