Learn More
Grainger Crimp Fitting 90 Deg 1/2 ID M24-1.5 F
Catalog No. 502763929
Encompass_Preferred
The Grainger 16P875 is a 3/4 HP, 115V, single phase, open drip proof motor with a 1075 RPM operating speed. It has a 48Y frame with a 1.15 service factor and features a cast iron body with standard foot mounting. The motor is thermally protected and suitable for continuous duty applications.
Catalog No. 50-276-3929 Supplier Grainger Supplier No. 16P875
The Grainger 16P875 is a 3/4 HP, 115V, single phase, open drip proof motor with a 1075 RPM operating speed. It has a 48Y frame with a 1.15 service factor and features a cast iron body with standard foot mounting. The motor is thermally protected and suitable for continuous duty applications.