Learn More
Grainger Crimp Fitting 90 Deg 1/4 ID 1/2 M
Catalog No. 502752689
Encompass_Preferred
The Grainger 14L762 is a Westward brand 1/4-inch drive socket set. It contains both SAE and metric sockets ranging from 4mm to 15mm and 1/8 inch to 9/16 inch. The set has a total of 38 pieces and comes in a molded case for storage and organization. The sockets are made from chrome vanadium steel for durability and have a matte finish to prevent corrosion. This socket set is ideal for automotive and machinery repair and maintenance.
Catalog No. 50-275-2689 Supplier Grainger Supplier No. 14L762
The Grainger 14L762 is a Westward brand 1/4-inch drive socket set. It contains both SAE and metric sockets ranging from 4mm to 15mm and 1/8 inch to 9/16 inch. The set has a total of 38 pieces and comes in a molded case for storage and organization. The sockets are made from chrome vanadium steel for durability and have a matte finish to prevent corrosion. This socket set is ideal for automotive and machinery repair and maintenance.