Learn More
Grainger Crimp Fitting 90 Deg 1-5/8 F JIC
Catalog No. 502752791
Encompass_Preferred
The DYE13977-20-20 is a 1-5/8" JIC 90 degree crimp fitting made by DYE. It is constructed of steel and has a maximum pressure rating of 5000 PSI. This fitting allows you to create a leak-proof 90 degree connection between two hydraulic hoses or tubes. It requires a hydraulic crimping machine to properly secure the hose or tube. This is an industrial grade fitting suitable for a variety of hydraulic applications.
Catalog No. 50-275-2791 Supplier Grainger Supplier No. 14L766
The DYE13977-20-20 is a 1-5/8" JIC 90 degree crimp fitting made by DYE. It is constructed of steel and has a maximum pressure rating of 5000 PSI. This fitting allows you to create a leak-proof 90 degree connection between two hydraulic hoses or tubes. It requires a hydraulic crimping machine to properly secure the hose or tube. This is an industrial grade fitting suitable for a variety of hydraulic applications.