Learn More
Grainger Crimp Fitting 90 Deg 1 ID M42-2 F
Catalog No. 502764708
Encompass_Preferred
The Grainger 16P878 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-scratching fibers for cleaning and scouring. It has a 6 inch by 9 inch size and comes in a pack of 10 pads. The pads are made in the USA and can be used for a variety of cleaning applications including pots, pans, dishes, walls, floors, grills and more. The open web design allows debris to move through the pad during use.
Catalog No. 50-276-4708 Supplier Grainger Supplier No. 16P878
The Grainger 16P878 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-scratching fibers for cleaning and scouring. It has a 6 inch by 9 inch size and comes in a pack of 10 pads. The pads are made in the USA and can be used for a variety of cleaning applications including pots, pans, dishes, walls, floors, grills and more. The open web design allows debris to move through the pad during use.