Learn More
Grainger Crimp Fitting 90 Deg 3/8 ID 3/8 FBSPP
Catalog No. 502766627
Encompass_Preferred
The Aeroquip 16P891 hydraulic crimp fitting is made of steel and has a 1/4" NPT male inlet and a #6 JIC 37° female outlet. It is rated for up to 5000 psi working pressure and 350°F operating temperature. The fitting has a zinc dichromate finish for corrosion resistance. It is designed for use in hydraulic systems to connect steel tubing to threaded ports. The crimp design provides a strong, reliable, vibration-resistant seal when used with the proper hose and crimp machine. This fitting allows flexible hose to interface with rigid plumbing in hydraulic machinery.
Catalog No. 50-276-6627 Supplier Grainger Supplier No. 16P795
The Aeroquip 16P891 hydraulic crimp fitting is made of steel and has a 1/4" NPT male inlet and a #6 JIC 37° female outlet. It is rated for up to 5000 psi working pressure and 350°F operating temperature. The fitting has a zinc dichromate finish for corrosion resistance. It is designed for use in hydraulic systems to connect steel tubing to threaded ports. The crimp design provides a strong, reliable, vibration-resistant seal when used with the proper hose and crimp machine. This fitting allows flexible hose to interface with rigid plumbing in hydraulic machinery.