Learn More
Grainger Crimp Fitting 90 Deg 5/8 I.D. M30-2 F
Catalog No. 502764687
Encompass_Preferred
The Grainger 16P876 is a 1/4 HP, 115V, 60Hz, 1-phase, 3450 RPM, 56C frame, open drip proof, continuous duty, C-face electric motor. It has a 48Y frame size with a 1.25" shaft diameter and 1.38" shaft height. The motor is thermally protected and rated for indoor use. It has ball bearings and meets NEMA design B specifications. The 16P876 has a full load current of 5.8 Amps and is suitable for operating pumps, fans, and other industrial equipment.
Catalog No. 50-276-4687 Supplier Grainger Supplier No. 16P876
The Grainger 16P876 is a 1/4 HP, 115V, 60Hz, 1-phase, 3450 RPM, 56C frame, open drip proof, continuous duty, C-face electric motor. It has a 48Y frame size with a 1.25" shaft diameter and 1.38" shaft height. The motor is thermally protected and rated for indoor use. It has ball bearings and meets NEMA design B specifications. The 16P876 has a full load current of 5.8 Amps and is suitable for operating pumps, fans, and other industrial equipment.