Learn More
Grainger Crimp Fitting 90 Deg 5/8 ID M26-1.5 F
Catalog No. 502764604
Encompass_Preferred
The Grainger Approved Crimp Fitting is a copper crimp fitting used for joining copper tubing. It has a 1/2" nominal pipe size and is made of wrought copper. The fitting has a pressure rating of 300 psi at 73°F and is NSF/ANSI 61 certified. It can be used for hot and cold potable water applications. The crimp fitting requires a crimping tool to secure the tubing. It provides a permanent, reliable connection and is ideal for residential and commercial plumbing and hydronic heating applications.
Catalog No. 50-276-4604 Supplier Grainger Supplier No. 16P839
The Grainger Approved Crimp Fitting is a copper crimp fitting used for joining copper tubing. It has a 1/2" nominal pipe size and is made of wrought copper. The fitting has a pressure rating of 300 psi at 73°F and is NSF/ANSI 61 certified. It can be used for hot and cold potable water applications. The crimp fitting requires a crimping tool to secure the tubing. It provides a permanent, reliable connection and is ideal for residential and commercial plumbing and hydronic heating applications.