Learn More
Chemglass Life Sciences Aluminum Seal, 11mm, Blue, PTFE/Silicone
SDP
Catalog No. 501532541
Encompass_Preferred
Aluminum Seal, 11mm, Blue, PTFE/Silicone. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. Pure Silicone laminated with .005 of PTFE. Recommended when multiple injections are required. Resist coring and excellent resealing qualities.
Catalog No. 50-153-2541 Supplier Chemglass Life Sciences Supplier No. CV3300B011
Aluminum Seal, 11mm, Blue, PTFE/Silicone. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. Pure Silicone laminated with .005 of PTFE. Recommended when multiple injections are required. Resist coring and excellent resealing qualities.