Learn More
Chemglass Life Sciences Aluminum Seal, 11mm, Magnetic, Silver, Red PTFE/White Silicone
SDP
Catalog No. 501532543
Encompass_Preferred
Aluminum Seal, 11mm, Magnetic, Silver, Red PTFE/White Silicone. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. The 11mm magnetic seal can be used with any instrument where the vials is transported with the use of a magnet.
Catalog No. 50-153-2543 Supplier Chemglass Life Sciences Supplier No. CV3300M011
Aluminum Seal, 11mm, Magnetic, Silver, Red PTFE/White Silicone. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. The 11mm magnetic seal can be used with any instrument where the vials is transported with the use of a magnet.