Learn More
Chemglass Life Sciences Aluminum Seal, 11mm, Red, PTFE/Silicone/PTFE
SDP
Catalog No. 501532548
Encompass_Preferred
Aluminum Seal, 11mm, Red, PTFE/Silicone/PTFE. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. Medium durometer silicone with .003 silicone laminated on both sides. Great for trace analysis work. Resist coring and good resealing characteristics. Use with auto samplers with blunt tip syringes.
Catalog No. 50-153-2548 Supplier Chemglass Life Sciences Supplier No. CV3400R011
Aluminum Seal, 11mm, Red, PTFE/Silicone/PTFE. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. Medium durometer silicone with .003 silicone laminated on both sides. Great for trace analysis work. Resist coring and good resealing characteristics. Use with auto samplers with blunt tip syringes.