Learn More
Chemglass Life Sciences Aluminum Seal, 11mm, Silver, Black Viton
SDP
Catalog No. 501532552
Encompass_Preferred
Aluminum Seal, 11mm, Silver, Black Viton. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. The seal with the viton septa is resistant to many aggressive solvents. Recommended for chlorinated solvents. Excellent chemical resistance.
Catalog No. 50-153-2552 Supplier Chemglass Life Sciences Supplier No. CV34500011
Aluminum Seal, 11mm, Silver, Black Viton. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. The seal with the viton septa is resistant to many aggressive solvents. Recommended for chlorinated solvents. Excellent chemical resistance.