Learn More
Chemglass Life Sciences Aluminum Seal, 11mm, Silver, PTFE/Silicone with Slit
SDP
Catalog No. 501532550
Encompass_Preferred
Aluminum Seal, 11mm, Silver, PTFE/Silicone with Slit. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. Pure Silicone laminated with .005 of PTFE. Recommended when multiple injections are required. Resist coring and excellent resealing qualities. Medium durometer silicone with .003 silicone laminated on both sides. Great for trace analysis work. Resist coring and good resealing characteristics. Use with auto samplers with blunt tip syringes.
Catalog No. 50-153-2550 Supplier Chemglass Life Sciences Supplier No. CV34100011
Aluminum Seal, 11mm, Silver, PTFE/Silicone with Slit. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. Pure Silicone laminated with .005 of PTFE. Recommended when multiple injections are required. Resist coring and excellent resealing qualities. Medium durometer silicone with .003 silicone laminated on both sides. Great for trace analysis work. Resist coring and good resealing characteristics. Use with auto samplers with blunt tip syringes.