Learn More
Chemglass Life Sciences Aluminum Seal, 11mm, Silver, Clear PTFE/Red Rubber
SDP
Catalog No. 50230934
Encompass_Preferred
Aluminum Seal, 11mm, Silver, Clear PTFE/Red Rubber. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. The seal with the viton septa is resistant to many aggressive solvents. The 11mm magnetic seal can be used with any instrument where the vials is transported with the use of a magnet.
Catalog No. 50-230-934 Supplier Chemglass Life Sciences Supplier No. CV31100011
Aluminum Seal, 11mm, Silver, Clear PTFE/Red Rubber. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. The seal with the viton septa is resistant to many aggressive solvents. The 11mm magnetic seal can be used with any instrument where the vials is transported with the use of a magnet.