Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Crimp seals with PTFE/Rubber Septa Seal

Catalog No. 111010303 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Product Type:
Pressure Release Crimp Cap
Diameter (Metric) Inner:
20 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Diameter (Metric) Inner
11-101-0303 Pressure Release Crimp Cap 20 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-0303 Supplier MilliporeSigma™ Supelco™ Supplier No. 27454U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Closure type crimp seal with rubber septum

  • PTFE/rubber septum
  • Crimp seals
  • Closure type
TRUSTED_SUSTAINABILITY

Specifications

Description Pressure release (Open center)
Diameter (Metric) Inner 20 mm
Color Silver
Material PTFE
Septum Rubber
No. per Pack 100
Quantity 100 Ea.
Thickness (Metric) 3.0 mm
Product Type Pressure Release Crimp Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.