Learn More
MilliporeSigma™ Supelco™ Crimp Seals with PTFE/Silicone Septa
Shop All MilliporeSigma Supelco ProductsDescription
- Used to seal the samples during the analysis of BTEX in BTEX contaminated waters, using static headspace extraction followed by gas chromatography with photoionization detection (HS-GC-PID).
Specifications
Specifications
| Description | Open center |
| Diameter (Metric) Opening | 10 mm |
| Color | Blue, Transparent, Silver |
| Material | Silicone, PTFE, Aluminum (Seal) |
| No. per Pack | 1000 |