Learn More
Qorpak™ Crimp Seals for Serum Vials
Catalog No. 02930157
Change view
Click to view available options
Quantity:
1000/Cs.
200/Cs.
5000/Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-930-157 1000/Cs.
02-930-156 200/Cs.
02-930-155 5000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-930-157 Supplier Qorpak™ Supplier No. 345416
Add to Cart
Add to Cart
Ensure the integrity and sterility of sensitive samples with our high-quality aluminum seals for serum vials
  • Reliable
  • Airtight
  • Tamper-evident
Specifications
Color Silver
Diameter (Metric) Outer 20 mm
Material Aluminum
Pre-slit No
Sterility Non-sterile
Quantity 1000/Cs.
Closure Size 20 mm
Autoclavable Autoclavable