Learn More
Bellco Glass, Inc. Crimp Top Amber Vial, 2.0mL 12x32mm - 11mm Crimp Top
SDP
Catalog No. 501945357
Encompass_Preferred
This 11mm crimp vial is made of amber borosilicate glass. Sample capacity of 2mL, 12mm in diameter, and 32mm in height. Vials compatible with 11mm crimp seals. 1,000 vials per case with 10 shelf packs of 100 vials packed in plastic shrink wrapped scallop packaging. MADE IN THE USA.
Catalog No. 50-194-5357 Supplier Bellco Glass, Inc. Supplier No. 207000040
This 11mm crimp vial is made of amber borosilicate glass. Sample capacity of 2mL, 12mm in diameter, and 32mm in height. Vials compatible with 11mm crimp seals. 1,000 vials per case with 10 shelf packs of 100 vials packed in plastic shrink wrapped scallop packaging. MADE IN THE USA.