Learn More
Bellco Glass, Inc. Crimp Top Clear Vial, 2.0mL 12x32mm - 11mm Crimp Top
SDP
Catalog No. 501945356
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Designed for use with Hewlett Packard and other auto-samplers. Large opening Vials have wider ID for easier sample access. Vials accept 11mm aluminum seals. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 12x32mmAluminum Seal Size: 11mmCase Quantity: 1000
Catalog No. 50-194-5356 Supplier Bellco Glass, Inc. Supplier No. 207000030
Manufactured from Type 1 Class A Borosilicate Glass. Designed for use with Hewlett Packard and other auto-samplers. Large opening Vials have wider ID for easier sample access. Vials accept 11mm aluminum seals. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 12x32mmAluminum Seal Size: 11mmCase Quantity: 1000