Learn More
World Precision Instrument Crimped Wire Mesh Sterilization Baskets, Tilted Handles-WP-4436CW
SDP
Catalog No. 502090343
Encompass
Stainless Steel Crimped Wire Mesh Sterilization Baskets with Tilt Handles. Electropolished and burr free to protect wraps and prevent injury. Meets all the requirements of automatic washing and sterilization for all types of instruments.
Catalog No. 50-209-0343 Supplier World Precision Instrument Supplier No. WP4436CW
Stainless Steel Crimped Wire Mesh Sterilization Baskets with Tilt Handles. Electropolished and burr free to protect wraps and prevent injury. Meets all the requirements of automatic washing and sterilization for all types of instruments.