Learn More
DWK Life Sciences Wheaton™ Crimper and Decapper Rechargeable Replacement Battery
SDP
Catalog No. 02915009 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
Crimpers and Decappers
Catalog No. For Use With (Equipment)
02-915-009 Crimpers and Decappers
1 options
Catalog No. 02-915-009 Supplier DWK Life Sciences Supplier No. W225700
Add to Cart
Add to Cart
Replacement Battery for Rechargeable Wheaton Crimper and Decappers
  • Lithium Ion Technology
  • Lasts up to 800 crimps per charge
  • Charges in one to two hour
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement Battery
For Use With (Equipment) Crimpers and Decappers
For Use With (Application) Rechafnging Battery Powered Crimpers and Decappers