Learn More
Microsolv Technology Corporation Crimping Tool, for 20mm, Pressure Relief, Aluminum Headspace Caps. Black handles. Hex adjustment key included. 1 EA.
Catalog No. 502269902
Encompass_Preferred
Chemical resistant coating makes these manual crimpers a great choice for laboratories. Adjustable height crimping jaws are made of a special, hardened alloy that produces a long life. The height of the crimp is easily adjusted by screwing up or down the pressure block in the crimping head with the supplied hex key. The maximum crimping pressure is controlled by the adjustable screw in the handle. Recommended for aluminum pressure relief (tear off seals) caps but not all steel crimp caps.
This item is not returnable. View return policy
Catalog No. 50-226-9902 Supplier Microsolv Technology Corporation Supplier No. 95025CRIMPF
Chemical resistant coating makes these manual crimpers a great choice for laboratories. Adjustable height crimping jaws are made of a special, hardened alloy that produces a long life. The height of the crimp is easily adjusted by screwing up or down the pressure block in the crimping head with the supplied hex key. The maximum crimping pressure is controlled by the adjustable screw in the handle. Recommended for aluminum pressure relief (tear off seals) caps but not all steel crimp caps.