Learn More
Medchemexpress LLC Crinecerfont hydroch 100mg | 321839-75-2 | 100MG
SDP
Catalog No. 5000243166
Encompass_Preferred
Crinecerfont (SSR-125543) hydrochloride is a potent orally active non-peptide CRF1 receptor antagonist Crinecerfont can be used for Classic congenital adrenal hyperplasia (CAH) research[1 Crinecerfont (hydrochloride) is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-002-43166 Supplier Medchemexpress LLC Supplier No. HY106203A100MG
Crinecerfont (SSR-125543) hydrochloride is a potent orally active non-peptide CRF1 receptor antagonist Crinecerfont can be used for Classic congenital adrenal hyperplasia (CAH) research[1 Crinecerfont (hydrochloride) is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups