Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Crinecerfont hydrochloride | 321839-75-2 | 98.0% | 519.50 | C27H29Cl2FN2OS | 5 MG
SDP

Catalog No. 5000242921
Encompass_Preferred

Crinecerfont hydrochloride is the hydrochloride salt of crinecerfont (SSR-125543), a potent, orally active, non-peptide corticotropin-releasing factor 1 (CRF1) receptor antagonist supplied for preclinical pharmacology research. It is provided as a high-purity solid with documented solubility and storage recommendations for laboratory use.

  • Potent CRF1 receptor antagonist for preclinical research.
  • High purity (98.0%).
  • Molecular weight 519.50 g/mol.
  • Soluble in DMSO at 100 mg/mL; in vivo formulation protocols available.
  • Available in small research quantities, such as 5 mg.
  • Store sealed away from moisture and light; follow cold storage guidance for solutions.

Catalog No. 50-002-42921 Supplier Medchemexpress LLC Supplier No. HY106203A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crinecerfont hydrochloride is the hydrochloride salt of crinecerfont (SSR-125543), a potent, orally active, non-peptide corticotropin-releasing factor 1 (CRF1) receptor antagonist supplied for preclinical pharmacology research. It is provided as a high-purity solid with documented solubility and storage recommendations for laboratory use.

  • Potent CRF1 receptor antagonist for preclinical research.
  • High purity (98.0%).
  • Molecular weight 519.50 g/mol.
  • Soluble in DMSO at 100 mg/mL; in vivo formulation protocols available.
  • Available in small research quantities, such as 5 mg.
  • Store sealed away from moisture and light; follow cold storage guidance for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.