Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Crinecerfont hydrochloride | 321839-75-2 | 98.0% | 519.50 g/mol | C27H29Cl2FN2OS | 10 MG
SDP

Catalog No. 5000244412
Encompass_Preferred

Crinecerfont hydrochloride is the hydrochloride salt of crinecerfont (SSR-125543), a potent, orally active, non-peptide corticotropin-releasing factor type 1 (CRF1) receptor antagonist supplied for laboratory research. It is provided as a light yellow to yellow solid with high purity and is intended for use in studies of CRF1-mediated pathways, including congenital adrenal hyperplasia research.

  • Potent CRF1 receptor antagonist, orally active for in vivo and in vitro studies.
  • High purity (98.0%) suitable for research applications.
  • Supplied as a light yellow to yellow solid for convenient handling.
  • Recommended storage conditions to maintain stability: solid at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Contains an alkyne group compatible with copper-catalyzed azide-alkyne cycloaddition for click chemistry applications.
  • Molecular weight 519.50 g/mol and molecular formula C27H29Cl2FN2OS.

Catalog No. 50-002-44412 Supplier Medchemexpress LLC Supplier No. HY106203A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crinecerfont hydrochloride is the hydrochloride salt of crinecerfont (SSR-125543), a potent, orally active, non-peptide corticotropin-releasing factor type 1 (CRF1) receptor antagonist supplied for laboratory research. It is provided as a light yellow to yellow solid with high purity and is intended for use in studies of CRF1-mediated pathways, including congenital adrenal hyperplasia research.

  • Potent CRF1 receptor antagonist, orally active for in vivo and in vitro studies.
  • High purity (98.0%) suitable for research applications.
  • Supplied as a light yellow to yellow solid for convenient handling.
  • Recommended storage conditions to maintain stability: solid at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Contains an alkyne group compatible with copper-catalyzed azide-alkyne cycloaddition for click chemistry applications.
  • Molecular weight 519.50 g/mol and molecular formula C27H29Cl2FN2OS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.