Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Crinecerfont hydrochloride | 321839-75-2 | MFCD05260970 | 98.0% | 519.50 g·mol⁻¹ | C27H29Cl2FN2OS | 50 MG
SDP

Catalog No. 5000243156
Encompass_Preferred

Crinecerfont hydrochloride is the hydrochloride salt of crinecerfont (SSR-125543), a selective corticotropin-releasing factor type-1 (CRF1) receptor antagonist supplied as a research reagent for pharmacology and preclinical studies.

  • Selective corticotropin-releasing factor type-1 (CRF1) receptor antagonist.
  • Intended for pharmacology and biochemical research applications.
  • Purity 98.0%.
  • Molecular weight 519.50 g·mol⁻¹.
  • Molecular formula C27H29Cl2FN2OS.
  • Typical packaging 50 mg.
  • CAS number 321839-75-2.

Catalog No. 50-002-43156 Supplier Medchemexpress LLC Supplier No. HY106203A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Crinecerfont hydrochloride is the hydrochloride salt of crinecerfont (SSR-125543), a selective corticotropin-releasing factor type-1 (CRF1) receptor antagonist supplied as a research reagent for pharmacology and preclinical studies.

  • Selective corticotropin-releasing factor type-1 (CRF1) receptor antagonist.
  • Intended for pharmacology and biochemical research applications.
  • Purity 98.0%.
  • Molecular weight 519.50 g·mol⁻¹.
  • Molecular formula C27H29Cl2FN2OS.
  • Typical packaging 50 mg.
  • CAS number 321839-75-2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.