Learn More
Sino Biological CRIP2 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 14552T56100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CRIP2 (rh CRIP2; Catalog#14552-H20B; P52943; Met1-Pro208). CRIP2 specific IgG was purified by Human CRIP2 affinity chromatography.
Catalog No. 50-224-8731