Learn More
TARGETMOL CHEMICALS INC Crisaborole 200MG
SDP
Catalog No. 502878769
Encompass_Preferred
Also available in 1 mL, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Crisaborole (AN-2728) is a phosphodiesterase 4 inhibitor. Purity 99.91%
Catalog No. 50-287-8769 Supplier TARGETMOL CHEMICALS INC Supplier No. T4285200MG
Also available in 1 mL, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Crisaborole (AN-2728) is a phosphodiesterase 4 inhibitor. Purity 99.91%