Learn More
Selleck Chemical LLC Crisaborole (AN2728) 25mg 906673-24-3
SDP
Supplier:  Selleck Chemical LLC S501425MG
Encompass_Preferred
Crisaborole (AN2728) is a small-molecule, boron-based, selective PDE4 inhibitor and has broad-spectrum anti-inflammatory activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8403