Learn More
Medchemexpress LLC Crisaborole-d4 | 2268785-42-6 | 98.8% | 255.07 | 1 ML
SDP
Catalog No. 5000345511
Encompass_Preferred
Crisaborole-d4 is deuterium labeled Crisaborole, a potent inhibitor of PDE4 and cytokine release. It inhibits PDE4 with an IC50 of 0.49 μM. This compound is suitable for use as a tracer and an internal standard for quantitative analysis, leveraging the potential of deuteration to affect pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Potent inhibitor of PDE4 and cytokine release
  • Suitable as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-45511 Supplier Medchemexpress LLC Supplier No. HY10978S1MG
Add to Cart
Add to Cart
Crisaborole-d4 is deuterium labeled Crisaborole, a potent inhibitor of PDE4 and cytokine release. It inhibits PDE4 with an IC50 of 0.49 μM. This compound is suitable for use as a tracer and an internal standard for quantitative analysis, leveraging the potential of deuteration to affect pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Potent inhibitor of PDE4 and cytokine release
  • Suitable as a tracer
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS