Learn More
Medchemexpress LLC Crisaborole (standard) | 906673-24-3 | 98.3% | 251.05 | 50 MG
SDP
Catalog No. 5000344958
Encompass_Preferred
Crisaborole (Standard) serves as an analytical standard for Crisaborole, a potent inhibitor of PDE4 and cytokine release with an IC50 of 0.49 μM. This reference standard is suitable for research and analytical applications.
  • Analytical standard for Crisaborole
  • Potent inhibitor of PDE4 and cytokine release
  • Used in qualitative, quantitative, and methodological research experiments
  • Applications include HPLC, GC, and MS
Catalog No. 50-003-44958 Supplier Medchemexpress LLC Supplier No. HY10978R10MG
Crisaborole (Standard) serves as an analytical standard for Crisaborole, a potent inhibitor of PDE4 and cytokine release with an IC50 of 0.49 μM. This reference standard is suitable for research and analytical applications.
  • Analytical standard for Crisaborole
  • Potent inhibitor of PDE4 and cytokine release
  • Used in qualitative, quantitative, and methodological research experiments
  • Applications include HPLC, GC, and MS