Learn More
Aviva Systems Biology Corp CRISPLD2 Antibody - N-terminal region (ARP50001_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50001_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRISPLD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CRISPLD2. Alternate names: CRISP11, DKFZP434B044, LCRISP2, MGC74865ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-60236