Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ CRISPR 3-D Model Kit
SCI_ED

Catalog No. S81449 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S81449 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81449 Supplier NewPath Learning™ Supplier No. 247717
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Assemble and use 3-D models to visualize and investigate key science structures.

  • Gain a critical understanding of complex structures & processes through the assembly of 3-dimensional models
  • Kit includes enough materials for students to assemble up to 10 paper models by working either independently or cooperatively in small groups
  • Also includes comprehensive Teacher Resource Guide outlining opportunities for your students to Engage, Explore, Explain, Extend & Evaluate their understanding of key Scientific concepts
  • Satisfies NGSS Standards which encourage students to develop & use models to represent structures, systems & processes, then provide explanations, make predictions or investigate solutions
  • Perfect for use in the classroom, lab or at home

Specifications

For Use With (Application) Model Assembly
Grade 6 to 10
Length (English) 12 in.
Material Paper
Weight (English) 2 lb.
Width (English) 9 in.
Product Type CRISPR
Grade Level Intermediate - Middle School, High School
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.