Learn More
Cellecta Inc Pooled Lentiviral CRISPR Human Genome 80K v2 Knockout Library, ~80,000 constructs, 4 sgRNAs per gene, storage -20°C (plasmid), -80°C (viral particles)
SDP
Catalog No. NC3718449
Encompass
Pooled Lentiviral CRISPR Human Genome 80K v2 Knockout Library, ~80,000 constructs, 4 sgRNAs per gene, storage -20°C (plasmid), -80°C (viral particles)
Catalog No. NC3718449 Supplier Cellecta Inc Supplier No. KOHGW80KV2V9
Add to Cart
Add to Cart
Request a Quote
Pooled Lentiviral CRISPR Human Genome 80K v2 Knockout Library, ~80,000 constructs, 4 sgRNAs per gene, storage -20°C (plasmid), -80°C (viral particles)