Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED CRISPRa Lentiviral dCas9-VPR Reagents, hEF1a, Viral Particles - 50µL, 10^7 TU/mL
Catalog No. NC1579853
Encompass
The CRISPR activation (CRISPRa) system is a variation of the canonical CRISPR-Cas9 used for creation of double-strand breaks in genomic DNA. It utilizes a nuclease-deactivated S. pyogenes Cas9 (dCas9), often called "dead Cas9", that is fused to one or more transcriptional activators. When paired with a well-designed guide RNA that targets a gene near a promoter region, the gene's native transcription start site is activated.
Catalog No. NC1579853 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. VCAS11922
The CRISPR activation (CRISPRa) system is a variation of the canonical CRISPR-Cas9 used for creation of double-strand breaks in genomic DNA. It utilizes a nuclease-deactivated S. pyogenes Cas9 (dCas9), often called "dead Cas9", that is fused to one or more transcriptional activators. When paired with a well-designed guide RNA that targets a gene near a promoter region, the gene's native transcription start site is activated.