Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED CRISPRMOD CRISPRA DCAS9-,100UG
Catalog No. NC2225184
Encompass
CRISPRmod CRISPRa dCas9-VPR mRNA, 100ugA DNA-free option for dCas9-VPR expressionPurified dCas9-VPR mRNA for co-transfection or electroporation with synthetic CRISPRa crRNA for gene activation
This item is not returnable. View return policy
Catalog No. NC2225184 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. CAS12212
CRISPRmod CRISPRa dCas9-VPR mRNA, 100ugA DNA-free option for dCas9-VPR expressionPurified dCas9-VPR mRNA for co-transfection or electroporation with synthetic CRISPRa crRNA for gene activation