Learn More
Bio Rad Laboratories CRIT GEL 4-20%TRIS18WELL,1.0MM
Catalog No. NC9753972
Encompass
Criterion Tris-HCl gels are precast polyacrylamide gels for vertical SDS-PAGE and native PAGE protein separations. Gels 13.3 cm x 8.7 cm x 1.0 mm (W x L x thickness) are supplied in a 15.0 x 10.6 cm cassette and are compatible with Criterio.
Catalog No. NC9753972 Supplier Bio Rad Laboratories Supplier No. 3450033
Criterion Tris-HCl gels are precast polyacrylamide gels for vertical SDS-PAGE and native PAGE protein separations. Gels 13.3 cm x 8.7 cm x 1.0 mm (W x L x thickness) are supplied in a 15.0 x 10.6 cm cassette and are compatible with Criterio.